Learn More
Strem, An Ascensus Company CAS# 32093-39-3. 5g. Hexakis(dimethylamino)dialuminum 98% (99.9%-Al) TDMAA.
SDP
Supplier:  Strem, An Ascensus Company 1345005G
Encompass
CAS# 32093-39-3. 5g. Hexakis(dimethylamino)dialuminum 98% (99.9%-Al) TDMAA. . Molecular Weight: 318.42. Molecular Formula: C12H36Al2N6. Color/form: white to yellow pwdr. Strem# 13-4500. http://www.strem.com/catalog/v/13-4500/
Catalog No. 50-204-5227