Learn More
Strem, An Ascensus Company CAS# 19782-68-4. 1g. Tetrakis(dimethylamino)hafnium, 98+% (99.99+%-Hf, <0.2% Zr) TDMAH, PURATREM. MFCD01862473
SDP
Supplier:  Strem, An Ascensus Company 7280001G
Encompass
CAS# 19782-68-4. 1g. Tetrakis(dimethylamino)hafnium, 98+% (99.99+%-Hf, <0.2% Zr) TDMAH, PURATREM. MFCD01862473. Molecular Weight: 354.79. Molecular Formula: Hf(N(CH3)2)4. Color/form: colorless to pale yellow xtl. Strem# 72-8000. http://www.strem.com/catalog/v/72-8000/
Catalog No. 50-201-6677