Learn More
Strem, An Ascensus Company CAS# [19782-68-4]. 25g. Tetrakis(dimethylamino)hafnium, 98+% (99.99+%-Hf, <0.2%-Zr) TDMAH, PURATREM, 72-8000, contained in 50 ml Swagelok cylinder (96-1070) for CVD/ALD. MFCD01862473
SDP
Supplier:  Strem, An Ascensus Company 98402125G
Encompass
CAS# 19782-68-4. 25g. Tetrakis(dimethylamino)hafnium, 98+% (99.99+%-Hf, <0.2%-Zr) TDMAH, PURATREM, 72-8000, contained in 50 ml Swagelok cylinder (96-1070) for CVD/ALD. MFCD01862473. Molecular Weight: 354.79. Molecular Formula: Hf(N(CH3)2)4. Color/form: colorless to pale yellow xtl. Strem# 98-4021. www.strem.com/catalog/v3/98-4021/
Catalog No. 50-197-9347