Learn More
Strem, An Ascensus Company CAS# 1066-77-9. 25g. Tetrakis(dimethylamino)tin(IV), 99% (99.99%-Sn) TDMASn PURATREM. MFCD00014860
SDP
Supplier:  Strem, An Ascensus Company 50181525G
Encompass
CAS# 1066-77-9. 25g. Tetrakis(dimethylamino)tin(IV), 99% (99.99%-Sn) TDMASn PURATREM. MFCD00014860. Molecular Weight: 295.01. Molecular Formula: Sn[N(CH3)2]4. Color/form: colorless to pale-yellow liq. Strem# 50-1815. http://www.strem.com/catalog/v/50-1815/
Catalog No. 50-201-6692