Learn More
Strem, An Ascensus Company CAS# [3275-24-9]. 25g. Tetrakis(dimethylamino)titanium(IV), 99% TDMAT. MFCD00014861
SDP
Catalog No. 501979344
Encompass
CAS# 3275-24-9. 25g. Tetrakis(dimethylamino)titanium(IV), 99% TDMAT. MFCD00014861. Molecular Weight: 224.20. Molecular Formula: Ti[N(CH3)2]4. Color/form: yellow to orange liq. Strem# 93-2240. www.strem.com/catalog/v3/93-2240/
Catalog No. 50-197-9344 Supplier Strem, An Ascensus Company Supplier No. 93224025G
CAS# 3275-24-9. 25g. Tetrakis(dimethylamino)titanium(IV), 99% TDMAT. MFCD00014861. Molecular Weight: 224.20. Molecular Formula: Ti[N(CH3)2]4. Color/form: yellow to orange liq. Strem# 93-2240. www.strem.com/catalog/v3/93-2240/