Learn More
Strem, An Ascensus Company CAS# [3275-24-9]. 5g. Tetrakis(dimethylamino)titanium(IV), 99% TDMAT. MFCD00014861
SDP
Catalog No. 501979345
Encompass
CAS# 3275-24-9. 5g. Tetrakis(dimethylamino)titanium(IV), 99% TDMAT. MFCD00014861. Molecular Weight: 224.20. Molecular Formula: Ti[N(CH3)2]4. Color/form: yellow to orange liq. Strem# 93-2240. www.strem.com/catalog/v3/93-2240/
Catalog No. 50-197-9345 Supplier Strem, An Ascensus Company Supplier No. 9322405G
CAS# 3275-24-9. 5g. Tetrakis(dimethylamino)titanium(IV), 99% TDMAT. MFCD00014861. Molecular Weight: 224.20. Molecular Formula: Ti[N(CH3)2]4. Color/form: yellow to orange liq. Strem# 93-2240. www.strem.com/catalog/v3/93-2240/