Learn More
Strem, An Ascensus Company CAS# [3275-24-9]. 5g. Tetrakis(dimethylamino)titanium(IV), 99% TDMAT (99.99%-Ti) PURATREM.
SDP
Catalog No. 501979365
Encompass
CAS# [3275-24-9]. 5g. Tetrakis(dimethylamino)titanium(IV), 99% TDMAT (99.99%-Ti) PURATREM. . Molecular Weight: 224.20. Molecular Formula: Ti[N(CH3)2]4. Color/form: yellow to orange liq. Strem# 22-2240. www.strem.com/catalog/v3/22-2240/
Catalog No. 50-197-9365 Supplier Strem, An Ascensus Company Supplier No. 2222405G
CAS# [3275-24-9]. 5g. Tetrakis(dimethylamino)titanium(IV), 99% TDMAT (99.99%-Ti) PURATREM. . Molecular Weight: 224.20. Molecular Formula: Ti[N(CH3)2]4. Color/form: yellow to orange liq. Strem# 22-2240. www.strem.com/catalog/v3/22-2240/