Learn More
Strem, An Ascensus Company CAS# [3275-24-9]. 25g. Tetrakis(dimethylamino)titanium(IV), 99% TDMAT, 93-2240, contained in 50 ml Swagelok cylinder (96-1070) for CVD/ALD. MFCD00014861
SDP
Catalog No. 501979369
Encompass
CAS# [3275-24-9]. 25g. Tetrakis(dimethylamino)titanium(IV), 99% TDMAT, 93-2240, contained in 50 ml Swagelok cylinder (96-1070) for CVD/ALD. MFCD00014861. Molecular Weight: 224.20. Molecular Formula: Ti[N(CH3)2]4. Color/form: yellow to orange liq. Strem# 98-4015. www.strem.com/catalog/v3/98-4015/
Catalog No. 50-197-9369 Supplier Strem, An Ascensus Company Supplier No. 98401525G
CAS# [3275-24-9]. 25g. Tetrakis(dimethylamino)titanium(IV), 99% TDMAT, 93-2240, contained in 50 ml Swagelok cylinder (96-1070) for CVD/ALD. MFCD00014861. Molecular Weight: 224.20. Molecular Formula: Ti[N(CH3)2]4. Color/form: yellow to orange liq. Strem# 98-4015. www.strem.com/catalog/v3/98-4015/