Learn More
Strem, An Ascensus Company CAS# 3275-24-9. 25g. Tetrakis(dimethylamino)titanium(IV), 99% TDMAT, 93-2240, contained in high-temp 50 ml Swagelok cylinder (96-1071) for CVD/ALD. MFCD00014861
SDP
Catalog No. 502045449
Encompass
CAS# 3275-24-9. 25g. Tetrakis(dimethylamino)titanium(IV), 99% TDMAT, 93-2240, contained in high-temp 50 ml Swagelok cylinder (96-1071) for CVD/ALD. MFCD00014861. Molecular Weight: 224.20. Molecular Formula: Ti[N(CH3)2]4. Color/form: yellow to orange liq. Strem# 98-4016. http://www.strem.com/catalog/v/98-4016/
Catalog No. 50-204-5449 Supplier Strem, An Ascensus Company Supplier No. 98401625G
CAS# 3275-24-9. 25g. Tetrakis(dimethylamino)titanium(IV), 99% TDMAT, 93-2240, contained in high-temp 50 ml Swagelok cylinder (96-1071) for CVD/ALD. MFCD00014861. Molecular Weight: 224.20. Molecular Formula: Ti[N(CH3)2]4. Color/form: yellow to orange liq. Strem# 98-4016. http://www.strem.com/catalog/v/98-4016/