Learn More
Strem, An Ascensus Company CAS# [19756-04-8]. 25g. Tetrakis(dimethylamino)zirconium(IV), 99% TDMAZ, 40-4100, contained in 50 ml Swagelok cylinder for (96-1070) CVD/ALD. MFCD00239502
SDP
Catalog No. 501979370
Encompass
CAS# [19756-04-8]. 25g. Tetrakis(dimethylamino)zirconium(IV), 99% TDMAZ, 40-4100, contained in 50 ml Swagelok cylinder for (96-1070) CVD/ALD. MFCD00239502. Molecular Weight: 267.53. Molecular Formula: Zr[N(CH3)2]4. Color/form: white to light yellow xtl. Strem# 98-4012. www.strem.com/catalog/v3/98-4012/
Catalog No. 50-197-9370 Supplier Strem, An Ascensus Company Supplier No. 98401225G
CAS# [19756-04-8]. 25g. Tetrakis(dimethylamino)zirconium(IV), 99% TDMAZ, 40-4100, contained in 50 ml Swagelok cylinder for (96-1070) CVD/ALD. MFCD00239502. Molecular Weight: 267.53. Molecular Formula: Zr[N(CH3)2]4. Color/form: white to light yellow xtl. Strem# 98-4012. www.strem.com/catalog/v3/98-4012/