Learn More
AdipoGen TDO human pAb
SDP
Supplier:  AdipoGen AG25A0106C100
Encompass_Preferred
Polyclonal Antibody. Recognizes human TDO. Detects a band of ~48kDa by Western blot. Source Rabbit. Applications ELISA, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. TDO incorporates oxygen into the indole moiety of tryptophan. Has a broad specificity towards tryptamine and derivatives including D- and L-tryptophan, 5-hydroxytryptophan and serotonin.
Catalog No. 50-256-3214