Learn More
Aviva Systems Biology Corp TDO2 Antibody - N-terminal region (ARP45716_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45716_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TDO2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human TDO2. Alternate names: TDO, TPH2, TRPO, TOApplicationIHC, WB. Species reactivity: Dog, Human, Pig
Catalog No. 50-014-59116