Learn More
Aviva Systems Biology Corp Tdp1 Antibody - middle region (ARP57199_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57199_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Tdp1. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 2810481F14Rik, 4921509N21Rik, AI838772, AW493413, E430034L06Rik, SCAN1, MLZ-501ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-276