Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 51567MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: TDP2/1258. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2b kappa. Positive Control: Jurkat cells. Lung, Duodenum or Small Intestinal Carcinoma. Cellular Staining: Nuclear ;Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human full-length TDP2 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 41kDa.Gene ID: 51567. Ensemble: ENSG00000111802. Uniprot: NP_057698.
Catalog No. 50-188-5042