Learn More
Aviva Systems Biology Corp Tdrd7 Antibody - C-terminal region (ARP55002_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55002_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Tdrd7. It was validated on Western Blot. . Alternate names: MGC93175, Pctaire2bpApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-988