Learn More
Aviva Systems Biology Corp TDRD9 Antibody - middle region (ARP36532_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36532_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TDRD9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TDRD9. Alternate names: C14orf75, DKFZp434N0820, FLJ36164, HIG-1, MGC135025, NET54ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56181