Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TDRL-551 10mM 1mL | 1644626-43-6 | 591.83 g·mol⁻1 | C25H23ClIN3O4 | 1 ML
SDP

Catalog No. 5000229298
Encompass_Preferred

Small-molecule replication protein A (RPA) inhibitor supplied as a 10 mM solution in DMSO (1 mL) for research use. Intended for in vitro studies of RPA-DNA interactions and combination effects with platinum-based chemotherapies.

  • Supplied as 10 mM solution in DMSO, 1 mL.
  • High purity (99.84%) suitable for biochemical assays.
  • Chemical formula C25H23ClIN3O4, molecular weight 591.83 g·mol⁻1.
  • Store frozen in solvent for stability (short- and long-term conditions apply).
  • Also available in multiple solid sizes for experimental flexibility.
  • For research use only; not for human or animal therapeutic use.

Catalog No. 50-002-29298 Supplier Medchemexpress LLC Supplier No. HY11484210MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Small-molecule replication protein A (RPA) inhibitor supplied as a 10 mM solution in DMSO (1 mL) for research use. Intended for in vitro studies of RPA-DNA interactions and combination effects with platinum-based chemotherapies.

  • Supplied as 10 mM solution in DMSO, 1 mL.
  • High purity (99.84%) suitable for biochemical assays.
  • Chemical formula C25H23ClIN3O4, molecular weight 591.83 g·mol⁻1.
  • Store frozen in solvent for stability (short- and long-term conditions apply).
  • Also available in multiple solid sizes for experimental flexibility.
  • For research use only; not for human or animal therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.