Learn More
Medchemexpress LLC TDSRCVIGLYHPPLQVY TFA | 99.9% | 2075.27 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP3460A10MG
Encompass_Preferred
TDSRCVIGLYHPPLQVY TFA is a disordered control peptide. It is a peptide containing the same amino acids as LP17 but in a different sequence order. This product is intended for research use only.
  • Disordered control peptide
  • Contains the same amino acids as LP17
  • Intended for research use only
Catalog No. 50-003-98700