Learn More
Genetex Inc TdT Monoclonal Antibody [MSVA-453R] HistoMAX™ (GTX04417) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04417
Encompass_Preferred
The TdT antibody [MSVA-453R] HistoMAX™ (GTX04417) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human Merkel-cell carcinoma from skin tissue using GTX04417 TdT antibody [MSVA-453R] HistoMAX. Merkel cell carcinoma with moderate to strong TdT immunostaining in > 80% of tumor cells.
Catalog No. NC3666895