Learn More
SUPERTECHS INC TDT CONTROL CELL SUSPENSION
Catalog No. NC9383813
Encompass
Supertechs Cat# 9500 is a fixed (paraformaldehyde) cell suspension containing human lymphoblastoid REH (TdT⁺) and RA-1 (TdT⁻) cells with stable TdT antigen. This cell suspension can be used as a control for TdT flow cytometric determinations. The control cell suspension contains a mixture of approximately 50% TdT⁺ and 50% TdT⁻ cells at a concentration of 2 x 10⁷ cells/mL (1.0 mL, ~20 tests) in a convenient dropper bottle. Our stabilized cell suspension has a shelf life of about 3 months after preparation when stored at 4°C. Control cells can be used for setting the threshold and gating in flow cytometry studies and for training technicians in performing and evaluating TdT flow cytometry experiments.
Catalog No. NC9383813 Supplier SUPERTECHS INC Supplier No. 9500
Supertechs Cat# 9500 is a fixed (paraformaldehyde) cell suspension containing human lymphoblastoid REH (TdT⁺) and RA-1 (TdT⁻) cells with stable TdT antigen. This cell suspension can be used as a control for TdT flow cytometric determinations. The control cell suspension contains a mixture of approximately 50% TdT⁺ and 50% TdT⁻ cells at a concentration of 2 x 10⁷ cells/mL (1.0 mL, ~20 tests) in a convenient dropper bottle. Our stabilized cell suspension has a shelf life of about 3 months after preparation when stored at 4°C. Control cells can be used for setting the threshold and gating in flow cytometry studies and for training technicians in performing and evaluating TdT flow cytometry experiments.