Learn More
Biocare Medical, LLC Terminal Deoxynucleotidyl Transferase Antibody - Predilute, 6.0 ml
SDP
Supplier:  Biocare Medical, LLC PP134AA
Encompass_Preferred
Terminal Deoxynucleotidyl Transferase antibody (TdT) is a nuclear protein widely used as a marker for lymphoblastic leukemia. TdT is a template-independent DNA polymerase which has been shown to be responsible for the addition of nucleotides at the N-region junction of rearranged Ig heavy chain and T-cell receptor gene segments during the maturation of B- and T-cells. Studies have shown that a panel of antibodies consisting of TdT, CD10, CD99 (MIC2), Bcl-2 and CD34 can be used to distinguish lymphoblastic leukemias from small noncleaved cell lymphomas.
Catalog No. 50-832-01