Learn More
ABclonal Technology TdT Rabbit mAb
SDP
Catalog No. 5000006914
Encompass_Preferred
This gene is a member of the DNA polymerase type-X family and encodes a template-independent DNA polymerase that catalyzes the addition of deoxynucleotides to the 3 -hydroxyl terminus of oligonucleotide primers In vivo the encoded protein is expressed in a restricted population of normal and malignant pre-B and pre-T lymphocytes during early differentiation where it generates antigen receptor diversity by synthesizing non-germ line elements (N-regions) at the junctions of rearranged Ig heavy chain and T cell receptor gene segments Alternatively spliced transcript variants encoding different isoforms of this gene have been described
Catalog No. 50-000-06914 Supplier ABclonal Technology Supplier No. A22618100UL
Add to Cart
Add to Cart
This gene is a member of the DNA polymerase type-X family and encodes a template-independent DNA polymerase that catalyzes the addition of deoxynucleotides to the 3 -hydroxyl terminus of oligonucleotide primers In vivo the encoded protein is expressed in a restricted population of normal and malignant pre-B and pre-T lymphocytes during early differentiation where it generates antigen receptor diversity by synthesizing non-germ line elements (N-regions) at the junctions of rearranged Ig heavy chain and T cell receptor gene segments Alternatively spliced transcript variants encoding different isoforms of this gene have been described