Learn More
Apexbio Technology LLC TDZD-8 327036-89-5 200mg
SDP
Catalog No. 5000088202
Encompass_Preferred
TDZD-8 (CAS 327036-89-5) is a selective non-ATP competitive inhibitor of glycogen synthase kinase 3 (GSK-3 ) exhibiting an IC50 of 1 4 M In addition to GSK-3 TDZD-8 also inhibits protein kinase C isoforms PKC I and PKC with IC50 values of 1 4 M and 1 1 M respectively Cellular studies have demonstrated that TDZD-8 suppresses both GSK-3 and PKC activity leading to reduced photodynamic therapy-induced neuronal necrosis and glial cell apoptosis Furthermore TDZD-8 has been shown to exert anti-leukemic effects in primary human leukemia cell models possibly through dual inhibition of PKC and FLT3 highlighting its utility in studies related to neuroprotection and leukemia
Catalog No. 50-000-88202 Supplier Apexbio Technology LLC Supplier No. B1249200
TDZD-8 (CAS 327036-89-5) is a selective non-ATP competitive inhibitor of glycogen synthase kinase 3 (GSK-3 ) exhibiting an IC50 of 1 4 M In addition to GSK-3 TDZD-8 also inhibits protein kinase C isoforms PKC I and PKC with IC50 values of 1 4 M and 1 1 M respectively Cellular studies have demonstrated that TDZD-8 suppresses both GSK-3 and PKC activity leading to reduced photodynamic therapy-induced neuronal necrosis and glial cell apoptosis Furthermore TDZD-8 has been shown to exert anti-leukemic effects in primary human leukemia cell models possibly through dual inhibition of PKC and FLT3 highlighting its utility in studies related to neuroprotection and leukemia