Learn More
TARGETMOL CHEMICALS INC TDZD-8 50MG
SDP
Catalog No. 502576580
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TDZD-8 (NP 01139) is an inhibitor of GSK-3(beta) with an IC50 of 2 uM; minimal inhibitory effect observed on CDK1 casein kinase II PKA and PKC. purity: 99%
Catalog No. 50-257-6580 Supplier TARGETMOL CHEMICALS INC Supplier No. T618750MG
Also available in 5 mg 10 mg 25 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TDZD-8 (NP 01139) is an inhibitor of GSK-3(beta) with an IC50 of 2 uM; minimal inhibitory effect observed on CDK1 casein kinase II PKA and PKC. purity: 99%