Learn More
Teknova TE Buffer (10mM Tris, 1mM EDTA), pH 8.0 Solution, 500mL. DNase/RNase Tested, PCR Grade.
SDP
Catalog No. NC9985640
Encompass
TE buffer is a salt solution that is commonly used to resuspend (or dilute) DNA and RNA. Composed of Tris base and EDTA, these molecules work together to maintain pH and prevent nucleic acids from degradation.
This item is not returnable. View return policy
Catalog No. NC9985640 Supplier Teknova Supplier No. T0228
TE buffer is a salt solution that is commonly used to resuspend (or dilute) DNA and RNA. Composed of Tris base and EDTA, these molecules work together to maintain pH and prevent nucleic acids from degradation.