Learn More
Bioworld TE Buffer (10X), 100 mL
Catalog No. 50196706
Encompass_Preferred
Product DescriptionTE (Tris-EDTA) has a liquid concentrate of 10X with a pH level of 8.0 brought up with HCl and is stored at room temperature. It contains 10 mM Tris and 1 mM EDTA. It is of biotechnology grade and no DNAse or RNAse activity is detected. TE buffer is used to solubilize DNA or RNA while protecting it from degradation. Commonly used for DNA isolation and electrophoresis protocols.
Catalog No. 50-196-706 Supplier Bioworld Supplier No. 105300291
Product DescriptionTE (Tris-EDTA) has a liquid concentrate of 10X with a pH level of 8.0 brought up with HCl and is stored at room temperature. It contains 10 mM Tris and 1 mM EDTA. It is of biotechnology grade and no DNAse or RNAse activity is detected. TE buffer is used to solubilize DNA or RNA while protecting it from degradation. Commonly used for DNA isolation and electrophoresis protocols.