Learn More
Research Products International Corp TE Buffer, 10X Powder, 158.35 Grams of Powder, Makes 10 Liters of Solution
SDP
Catalog No. 50213690
Encompass_Preferred
Dry homogeneous blend of molecular biology grade Tris-base and EDTA, disodium salt. For use in gel electrophoresis.
Supplied as 158.35 grams of powder which is used to produce a 10X concentrate.
Catalog No. 50-213-690 Supplier Research Products International Corp Supplier No. T2204010.0
Dry homogeneous blend of molecular biology grade Tris-base and EDTA, disodium salt. For use in gel electrophoresis.
Supplied as 158.35 grams of powder which is used to produce a 10X concentrate.