Learn More
Teknova TE Buffer (10mM Tris, 1mM EDTA), pH 8.0 Solution. 1000 ml DNAse/RNAse Free.
SDP
Catalog No. 50843206
Encompass
TE buffer is a salt solution that is commonly used to resuspend (or dilute) DNA and RNA. Composed of Tris base and EDTA, these molecules work together to maintain pH and prevent nucleic acids from degradation.
This item is not returnable. View return policy
Catalog No. 50-843-206 Supplier Teknova Supplier No. T0226
TE buffer is a salt solution that is commonly used to resuspend (or dilute) DNA and RNA. Composed of Tris base and EDTA, these molecules work together to maintain pH and prevent nucleic acids from degradation.