Learn More
Boston Bioproducts Inc Tris-EDTA Buffer, TE (20X, pH 8.0) - 250ml
Catalog No. NC9217211
Encompass
The Tris-EDTA (TE) Buffer is a commonly used buffer solution in molecular biology, especially in processes involving nucleic acids. This buffer is composed of a 20X concentration of Tris, a buffering agent, and EDTA, which has the ability to prevent degradation of DNA and RNA by chelating magnesium or other divalent metal ions, and allows pH regulation within the range of 7.5-8.5. It is widely used often for DNA and RNA gel electrophoresis, and also for DNA automated sequencing.
Catalog No. NC9217211 Supplier Boston Bioproducts Inc Supplier No. BM304250ML
The Tris-EDTA (TE) Buffer is a commonly used buffer solution in molecular biology, especially in processes involving nucleic acids. This buffer is composed of a 20X concentration of Tris, a buffering agent, and EDTA, which has the ability to prevent degradation of DNA and RNA by chelating magnesium or other divalent metal ions, and allows pH regulation within the range of 7.5-8.5. It is widely used often for DNA and RNA gel electrophoresis, and also for DNA automated sequencing.