Learn More
Research Products International Corp TE Buffer 20X Solution pH 8.0, 250 Milliliters
SDP
Catalog No. 50997996
Encompass_Preferred
Formulation:
TRIS HCI: 200 mM
EDTA: 20 mM
Catalog No. 50-997-996 Supplier Research Products International Corp Supplier No. T32200250.0
Formulation:
TRIS HCI: 200 mM
EDTA: 20 mM