Learn More
LGC GENOMICS LLC (Lucigen) TE Buffer, 70 mLStandard buffer for storing nucleic acids (10 mM Tris-HCl [pH 7.5], 1 mM EDTA). To be used with the MasterPure Kits and any other nucleic acid extraction/purification kits.
Catalog No. NC9331805
Encompass
This reagent is designed for use with the MasterPure™ kit protocols only.
This item is not returnable. View return policy
Catalog No. NC9331805 Supplier LGC GENOMICS LLC Supplier No. MTE0970
This reagent is designed for use with the MasterPure™ kit protocols only.