Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TE buffer, pH 7.4, RNAse free

Catalog No. AAJ60234AK
Change view
Click to view available options
Quantity:
125 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60234AK 250 mL
AAJ60234EQE 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60234AK Supplier Thermo Scientific Chemicals Supplier No. J60234AK
  • $171.00
    $124.65 / Each of 1

    Save $46.35

    27% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1185-53-1 | C4H12ClNO3 | 157.594 g/mol

TE buffer is a commonly used buffer solution in procedures involving DNA or RNA. It is used for storing and diluting DNA and RNA. Treatment with TE buffer enhances the staining intensity of antibodies in the immunohistochemical detection of certain proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
TE buffer is a commonly used buffer solution in procedures involving DNA or RNA. It is used for storing and diluting DNA and RNA. Treatment with TE buffer enhances the staining intensity of antibodies in the immunohistochemical detection of certain proteins.

Solubility
Fully miscible in water.

Notes
200mM Tris-HCl with 20mM EDTA
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1185-53-1
Molecular Formula C4H12ClNO3
Molecular Weight (g/mol) 157.594
MDL Number MFCD00236359
InChI Key QKNYBSVHEMOAJP-UHFFFAOYSA-N
PubChem CID 93573
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
SMILES C(C(CO)(CO)N)O.Cl

Specifications

Color Undesignated
pH 7.4
Physical Form Liquid
Quantity 250 mL
Solubility Information Fully miscible in water.
Chemical Name or Material TE buffer, RNAse free
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.