Learn More
Teknova DNA Suspension Buffer, pH 8.0. RNase/DNase Tested, PCR. 250mL, Sterile. (10mM Tris, 0.1mM EDTA)
SDP
Catalog No. 50843203
Encompass
TE buffer is a salt solution that is commonly used to resuspend (or dilute) DNA and RNA. Composed of Tris base and EDTA, these molecules work together to maintain pH and prevent nucleic acids from degradation.
This item is not returnable. View return policy
Catalog No. 50-843-203 Supplier Teknova Supplier No. T0223
Add to Cart
Add to Cart
TE buffer is a salt solution that is commonly used to resuspend (or dilute) DNA and RNA. Composed of Tris base and EDTA, these molecules work together to maintain pH and prevent nucleic acids from degradation.