Learn More
Specifications
Specifications
| Chemical Name or Material | TE Buffer Solution |
| CAS | 77-86-1, 6381-92-6, 7647-01-0, 1310-73-2 |
| For Use With (Application) | Molecular Biology |
| Physical Form | Aqueous Solution |
| Quantity | 100 mL |
| Solution Type | Buffer, Storage |
| DNase | None Detected |
| Protease | None Detected |
| Solubility Information | Soluble in water |
| Grade | Molecular Biology |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.