Learn More
MSC TE-CO Tapped Hole Location Gage, 4-40 UNC, 52100 Steel Body, 12L14 Steel Collar
Catalog No. 5000530174
Encompass
This Tapped Hole Location Gage is designed for accurately measuring the distance between two tapped holes in a workpiece. It features a robust two-piece construction with high-quality steel components, ensuring precision and durability for various applications.
  • Body constructed from 52100 steel
  • Collar made from 12L14 steel
  • Black oxide finish applied to the collar
  • Two-piece design for reliable performance
  • Consistent shaft size for simplified hole spread calculations
  • All diameters are concentric within 0.0002 T.I.R.
  • Available with English (Thread Class: 3A) and Metric (Thread Class: 6G) threading options
Catalog No. 50-005-30174 Supplier MSC Supplier No. 84343995
This Tapped Hole Location Gage is designed for accurately measuring the distance between two tapped holes in a workpiece. It features a robust two-piece construction with high-quality steel components, ensuring precision and durability for various applications.
  • Body constructed from 52100 steel
  • Collar made from 12L14 steel
  • Black oxide finish applied to the collar
  • Two-piece design for reliable performance
  • Consistent shaft size for simplified hole spread calculations
  • All diameters are concentric within 0.0002 T.I.R.
  • Available with English (Thread Class: 3A) and Metric (Thread Class: 6G) threading options