Learn More
Quality Biological Inc TE pH 7.4, 1000 mL
SDP
Catalog No. 507517264
Encompass_Preferred
TE is composed of buffering agent Tris, and chelating agent EDTA. ¿ EDTA prevents degradation of DNA and RNA by chelating magnesium (or other divalent metal ions). ¿ Together, this Tris-EDTA based solution solubilizes DNA or RNA, while protecting it from degradation. ¿ Applications: TE is useful for reconstituting nucleic acids, during DNA precipitation and other DNA applications, storing and diluting DNA and RNA, breaking protein-links in immuno-histochemistry procedures, and unmasking antigens and epitopes in formalin-fixed and paraffin-embedded tissue sections. ¿ Storage Conditions: 15¿C to 30¿C ¿ Shipping Condition: ambient temperature ¿ Sterile Filtered ¿ DNase, RNase and Protease Tested ¿ Product and/or packaging contains recycled/renewable content ¿ Product and/or packaging is recyclable or reduces waste ¿ Product and/or packaging is safer to human and environmental health ¿ ¿ Max Shelf Life: 36 months
Catalog No. 50-751-7264 Supplier Quality Biological Inc Supplier No. 351010131
Add to Cart
Add to Cart
TE is composed of buffering agent Tris, and chelating agent EDTA. ¿ EDTA prevents degradation of DNA and RNA by chelating magnesium (or other divalent metal ions). ¿ Together, this Tris-EDTA based solution solubilizes DNA or RNA, while protecting it from degradation. ¿ Applications: TE is useful for reconstituting nucleic acids, during DNA precipitation and other DNA applications, storing and diluting DNA and RNA, breaking protein-links in immuno-histochemistry procedures, and unmasking antigens and epitopes in formalin-fixed and paraffin-embedded tissue sections. ¿ Storage Conditions: 15¿C to 30¿C ¿ Shipping Condition: ambient temperature ¿ Sterile Filtered ¿ DNase, RNase and Protease Tested ¿ Product and/or packaging contains recycled/renewable content ¿ Product and/or packaging is recyclable or reduces waste ¿ Product and/or packaging is safer to human and environmental health ¿ ¿ Max Shelf Life: 36 months