Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc TE, pH 7.4 100mL
SDP

Catalog No. 502209772
Encompass_Preferred

TE is composed of buffering agent Tris, and chelating agent EDTA. EDTA prevents degradation of DNA and RNA by chelating magnesium (or other divalent metal ions). Together, this Tris-EDTA based solution solubilizes DNA or RNA, while protecting it from degradation.

Applications: TE is useful for reconstituting nucleic acids, during DNA precipitation and other DNA applications, storing and diluting DNA and RNA, breaking protein-links in immuno-histochemistry procedures, and unmasking antigens and epitopes in formalin-fixed and paraffin-embedded tissue sections.

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered
DNase, RNase and Protease Tested
Max Shelf Life: 36 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. 50-220-9772 Supplier Quality Biological Inc Supplier No. 351010721
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TE is composed of buffering agent Tris, and chelating agent EDTA. EDTA prevents degradation of DNA and RNA by chelating magnesium (or other divalent metal ions). Together, this Tris-EDTA based solution solubilizes DNA or RNA, while protecting it from degradation.

Applications: TE is useful for reconstituting nucleic acids, during DNA precipitation and other DNA applications, storing and diluting DNA and RNA, breaking protein-links in immuno-histochemistry procedures, and unmasking antigens and epitopes in formalin-fixed and paraffin-embedded tissue sections.

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered
DNase, RNase and Protease Tested
Max Shelf Life: 36 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.