Learn More
Active Motif TE, pH 8.0
SDP
Catalog No. NC1847628
Encompass
Active Motif’s TE, pH 8.0 is a commonly used buffer solution in molecular biology. TE buffer can be used for a variety of applications including solubility of DNA or RNA solutions. TE buffer is also used to wash Protein G agarose beads prior to use in immunoprecipitation. TE buffer is 10 mM Tris-HCl, 1 mM EDTA, pH 8.0 and is provided ready to use.
Catalog No. NC1847628 Supplier Active Motif Supplier No. 37515
Add to Cart
Edge
Add to Cart
Active Motif’s TE, pH 8.0 is a commonly used buffer solution in molecular biology. TE buffer can be used for a variety of applications including solubility of DNA or RNA solutions. TE buffer is also used to wash Protein G agarose beads prior to use in immunoprecipitation. TE buffer is 10 mM Tris-HCl, 1 mM EDTA, pH 8.0 and is provided ready to use.