Learn More
Chemglass Life Sciences Stopcock, Double Oblique Bore, PTFE Plug, 2mm Bore, 12.9/44 Plug Size, 8mm OD Sidearms
SDP
Catalog No. CG44602
Encompass_Preferred
Stopcock, Double Oblique Bore, PTFE Plug, 2mm Bore, 12.9/44 Plug Size, 8mm OD Sidearms. A 1:5 taper PTFE stopcock with two parallel diagonal bores.
Catalog No. CG44602 Supplier Chemglass Life Sciences Supplier No. CG44602
Stopcock, Double Oblique Bore, PTFE Plug, 2mm Bore, 12.9/44 Plug Size, 8mm OD Sidearms. A 1:5 taper PTFE stopcock with two parallel diagonal bores.