Quickly and efficiently transfers proteins and nucleic acids from small gels to nylon, nitrocellulose or PVDF membranesFeatures and BenefitsTransfer as many as four small gels, up to 9 x 10 cm in less than an hourUniform and strong electric field—supports efficient and even transfersColor-coded, easy to assemble and load cassettes—ensures proper orientation during transferSuperior tank design allows the cassettes to apply equal pressure across the stack—prevents gel distortionBuilt-in ceramic heat exchanger—provides excellent temperature control when used with an external cooling water bathBuilt-in alumina-covered cooling channel provides excellent temperature control with no more than a 5°C temperature increase during a typical runIncludes:Lower Chamber with Heat Exchanger; Safety Lid with High Voltage Leads; Electrode Panels, 2 pcs; Cassettes, 4 pcs; Foam Sponges, 6 mm thick, 4 pcs; Foam Sponges, 3 mm thick, 8 pcs; Cassette Hook; Blotter Paper (9 x 10.5 cm), 25 sheets