Learn More
Julabo USA Inc TE400 EA 100-230V/50-60Hz
SDP
Catalog No. NC2747044
Encompass_Preferred
Peltier circulator, JULABO, TE400 Peltier chiller 100-230V/50-60Hz, No refrigerant; -5…+40 °C; 450kW heater. Cooling: 400W at 20C; 200W at 10C. Pressure pump 7L/min, 17.43PSI. Color touchscreen UI with RS232, multi-lingual, programmable. M16x1 pump fittings 8 and 12mm barbs. 1L bath tank. 10.8 x 16.3 x 15.7 inhes (WxLxH), 53lbs
Catalog No. NC2747044 Supplier Julabo USA Inc Supplier No. 9635400.99
Add to Cart
Add to Cart
Request a Quote
Peltier circulator, JULABO, TE400 Peltier chiller 100-230V/50-60Hz, No refrigerant; -5…+40 °C; 450kW heater. Cooling: 400W at 20C; 200W at 10C. Pressure pump 7L/min, 17.43PSI. Color touchscreen UI with RS232, multi-lingual, programmable. M16x1 pump fittings 8 and 12mm barbs. 1L bath tank. 10.8 x 16.3 x 15.7 inhes (WxLxH), 53lbs