Learn More
Hoefer, Inc Hoefer™ TE42 Standard Transfer Tank
SDP
Catalog No. NC2740688
Encompass_Preferred
Quickly and evenly transfers proteins and nucleic acids from polyacrylamide or agarose gels onto nylon, nitrocellulose, or PVDF membranesFeatures & Benefits:Uniform and strong electric field—supports efficient and even transfersColor-coded, easy to assemble cassettes—ensures proper orientation during transferSuperior tank design allows the cassettes to apply equal pressure across the stack—prevents gel distortionOptional TE47 heat exchanger—provides excellent buffer temperature control when used with an external cooling water bathIncludes:Lower Chamber; Safety Lid with High Voltage Leads; Electrode Panels, 2 pcs; Cassettes, 2 pcs; Foam Sponges, 6 mm thick, 2 pcs; Foam Sponges, 3 mm thick, 4 pcs; Cassette Hook; Blotter Paper (14.5 x 21.5 cm), 25 sheetsSpecifications:Capacity: Up to four 15 x 21 cm gels (without cooling), Up to two 15 x 21 cm gels (with cooling)Maximum Power Settings 100 V, 2 A, 200 WUnit Dimensions (W x H x D) 8 x 13 x 30.5 cm
Catalog No. NC2740688 Supplier Hoefer, Inc Supplier No. TE42
Quickly and evenly transfers proteins and nucleic acids from polyacrylamide or agarose gels onto nylon, nitrocellulose, or PVDF membranesFeatures & Benefits:Uniform and strong electric field—supports efficient and even transfersColor-coded, easy to assemble cassettes—ensures proper orientation during transferSuperior tank design allows the cassettes to apply equal pressure across the stack—prevents gel distortionOptional TE47 heat exchanger—provides excellent buffer temperature control when used with an external cooling water bathIncludes:Lower Chamber; Safety Lid with High Voltage Leads; Electrode Panels, 2 pcs; Cassettes, 2 pcs; Foam Sponges, 6 mm thick, 2 pcs; Foam Sponges, 3 mm thick, 4 pcs; Cassette Hook; Blotter Paper (14.5 x 21.5 cm), 25 sheetsSpecifications:Capacity: Up to four 15 x 21 cm gels (without cooling), Up to two 15 x 21 cm gels (with cooling)Maximum Power Settings 100 V, 2 A, 200 WUnit Dimensions (W x H x D) 8 x 13 x 30.5 cm