Learn More
Hoefer, Inc TE62 Standard Transfer Tank with Cooling Chamber
SDP
Catalog No. NC2743277
Encompass_Preferred
Quickly and evenly transfers proteins and nucleic acids from polyacrylamide or agarose gels onto nylon, nitrocellulose, or PVDF membranes while providing excellent temperature controlThe TE62 is designed to accept up to four cassettes simultaneously with cooling. Each cassette holds a 15 x 21 cm gel, or as many as four 7 x 10 cm mini gels.Features & Benefits:Uniform and strong electric field—supports efficient and even transfersColor-coded, easy to assemble cassettes—ensures proper orientation during transferSuperior tank design allows the cassettes to apply equal pressure across the stack—prevents gel distortionBuilt-in ceramic heat exchanger—provides excellent temperature control when used with an external cooling water bath Includes:Lower Chamber with Heat Exchanger; Safety Lid with High Voltage Leads; Electrode Panels, 2 pcs; Cassettes, 4 pcs; Foam Sponges, 6 mm thick, 4 pcs; Foam Sponges, 3 mm thick, 8 pcs; Cassette Hook; Blotter Paper (14.5 x 21.5 cm), 25 sheets
Catalog No. NC2743277 Supplier Hoefer, Inc Supplier No. TE62
Quickly and evenly transfers proteins and nucleic acids from polyacrylamide or agarose gels onto nylon, nitrocellulose, or PVDF membranes while providing excellent temperature controlThe TE62 is designed to accept up to four cassettes simultaneously with cooling. Each cassette holds a 15 x 21 cm gel, or as many as four 7 x 10 cm mini gels.Features & Benefits:Uniform and strong electric field—supports efficient and even transfersColor-coded, easy to assemble cassettes—ensures proper orientation during transferSuperior tank design allows the cassettes to apply equal pressure across the stack—prevents gel distortionBuilt-in ceramic heat exchanger—provides excellent temperature control when used with an external cooling water bath Includes:Lower Chamber with Heat Exchanger; Safety Lid with High Voltage Leads; Electrode Panels, 2 pcs; Cassettes, 4 pcs; Foam Sponges, 6 mm thick, 4 pcs; Foam Sponges, 3 mm thick, 8 pcs; Cassette Hook; Blotter Paper (14.5 x 21.5 cm), 25 sheets