Learn More
Hoefer, Inc Hoefer™ TE77X Semi-Dry Transfer Units
SDP
Catalog No. NC2740716
Encompass_Preferred
Efficient design that uses minimal buffer to transfer proteins from polyacrylamide gels in less than an hourThe Semi-Dry Transfer Units support as many as two layers of gels being transferred simultaneously. The TE77X has a 21 x 26 cm transfer area for a capacity of up to 12 mini gels at one time.Features & Benefits:Included safety circuit breaker—limits voltage and current from the user's power supply, preventing electrical damage to the transfer unitRequires minimal current—does not generate the excessive heat that can dry out the transfer stack and halt transfer or damage transfer units Durable iridium oxide and stainless steel electrodes—allow for contamination free, consistent transferMinimal buffer requirements—reagent cost and preparation time are reducedIncludes:Molded Base with Platinum-Coated; Titanium Anode and High Voltage Leads; Hinged Lid with Stainless Steel Cathode; Blotting Paper (21 x 26 cm); Porous Cellophane (20 x 35.5 cm); Mylar Masks (23 x 27.5 cm)
Catalog No. NC2740716 Supplier Hoefer, Inc Supplier No. TE77X
Efficient design that uses minimal buffer to transfer proteins from polyacrylamide gels in less than an hourThe Semi-Dry Transfer Units support as many as two layers of gels being transferred simultaneously. The TE77X has a 21 x 26 cm transfer area for a capacity of up to 12 mini gels at one time.Features & Benefits:Included safety circuit breaker—limits voltage and current from the user's power supply, preventing electrical damage to the transfer unitRequires minimal current—does not generate the excessive heat that can dry out the transfer stack and halt transfer or damage transfer units Durable iridium oxide and stainless steel electrodes—allow for contamination free, consistent transferMinimal buffer requirements—reagent cost and preparation time are reducedIncludes:Molded Base with Platinum-Coated; Titanium Anode and High Voltage Leads; Hinged Lid with Stainless Steel Cathode; Blotting Paper (21 x 26 cm); Porous Cellophane (20 x 35.5 cm); Mylar Masks (23 x 27.5 cm)