Learn More
American Radiolabeled Chemicals Inc TEA CL-1-14C 250UC
SDP
Catalog No. 5090106681
Encompass_Preferred
Tetraethylammonium chloride-ethyl-1-14C 250uCi in ethanol, 0.1 mCi/ml, S.A. 50-60 mCi/mmol, M.W. 151.0, Exclusive
Catalog No. 50-901-06681 Supplier American Radiolabeled Chemicals Inc Supplier No. ARC 1540250UC
Add to Cart
Add to Cart
Request a Quote
Tetraethylammonium chloride-ethyl-1-14C 250uCi in ethanol, 0.1 mCi/ml, S.A. 50-60 mCi/mmol, M.W. 151.0, Exclusive