Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tea Extract from Camellia sinensis, ≥80% theaflavins (theaflavin and theaflavin gallates) basis, MilliporeSigma™ Supelco™

Catalog No. 111005198 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-5198 1 mg
11-100-4343 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-5198 Supplier MilliporeSigma Supelco Supplier No. T55501MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Featured Industries: Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

Physical Form Neat
Type Analytical Standard
Quantity 1 mg
Percent Purity ≥80% theaflavins (theaflavin and theaflavin gallates) basis
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.