Learn More
Metrohm USA TEABR REF SOLN 6 2320.000TEABR
SDP
Supplier:  Metrohm USA 62320000
Encompass
Electrolyte solution TEABr (tetraethylammonium bromide in ethylene glycol), c(TEABr) = 0.4 mol/L
Catalog No. NC0205174