Learn More
National Optical Teaching Laboratory Biological Advanced Microscope with Camera
SCI_ED
Catalog No. S04981
Change view
Click to view available options
Head Style:
Trinocular
Catalog No. Head Style
S04981 Trinocular
1 options
Catalog No. S04981 Supplier National Optical Supplier No. DSTELLAR 1 PROT4X5
The Stellar Series teaching and laboratory biological microscopes adopt a new structural design, with a smoother appearance and stable performance.
  • The combination of a Stellar Pro Series teaching biological microscope with 100X oil immersion lens and digital solution enables you to display and share live images.
  • Discover the possibilities and benefits of adding an integrated tablet or microscope camera to your Pro Series microscope.
  • Choose from integrated 8 inch Android-based tablet or WiFi enabled microscope camera.
Specifications
Camera Type WiFi Enabled
Eyepiece WF 10X/18 mm
Head Style Trinocular
Lighting Type LED
Magnification Power 4X, 10X, 40X, 100X
Model Stellar
Objective Types Infinity Corrected
Product Type Biological Microscope
Includes Camera
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov