Learn More
National Optical Teaching Laboratory Biological Microscope
SCI_ED
Catalog No. S04979
Change view
Click to view available options
Head Style:
Binocular
Trinocular
Magnification Power:
4X, 10X, 40X
4X, 10X, 40X, 100X
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Head Style Magnification Power
S04979 Trinocular 4X, 10X, 40X, 100X
S04988 Binocular 4X, 10X, 40X
S04987 Binocular 4X, 10X, 40X, 100X
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S04979 Supplier National Optical Supplier No. DSTELLAR 1 T4
Only null left
Add to Cart
Add to Cart
The Stellar Series teaching and laboratory biological microscopes adopt a new structural design, with a smoother appearance and stable performance.
  • The Stellar Series teaching biological microscopes adopts a new structural design and updated appearance.
  • This microscope comes with DIN Achromat objectives, mechanical stage, Coaxial adjustment, built in cord wrap, built in slide holder and LED illumination.
  • Choose between binocular or trinocular heads with a choice of three objectives (4X, 10X, 40X) or four objectives which includes the 100X oil immersion lens.
Specifications
Eyepiece WF 10X/18 mm
Head Style Trinocular
Lighting Type LED
Magnification Power 4X, 10X, 40X, 100X
Model Stellar
Objective Types Achromatic
Product Type Biological Microscope
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov