Learn More
World Precision Instrument Teaching Microscope System
SDP
Catalog No. 502538577
Encompass
Share what you see under the microscope with a group audience
Looking for a way to share what you see under the microscope with a remote audience? The WPI SMARTPZM Teaching Microscope System is perfect for remote learning or even social distancing in a live classroom. The HD color microscopy cameras set a new standard for excellence in high definition imaging for scientific and industrial applications. The HD cameras are full-featured offering 30 fps in video preview mode, with unrivaled color fidelity and on-board image capturing capability. 
Catalog No. 50-253-8577 Supplier World Precision Instrument Supplier No. SMARTPZM
Add to Cart
Add to Cart
Request a Quote
Share what you see under the microscope with a group audience
Looking for a way to share what you see under the microscope with a remote audience? The WPI SMARTPZM Teaching Microscope System is perfect for remote learning or even social distancing in a live classroom. The HD color microscopy cameras set a new standard for excellence in high definition imaging for scientific and industrial applications. The HD cameras are full-featured offering 30 fps in video preview mode, with unrivaled color fidelity and on-board image capturing capability. 